Learn More
Description
Specifications
Specifications
| Antigen | FAM125A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | CFBP, CIN85/CD2AP family binding protein, CIN85/CD2AP family-binding protein, ESCRT-I complex subunit MVB12A, family with sequence similarity 125, member A, FLJ32495, multivesicular body subunit 12A, Protein FAM125A |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human FAM125A (NP_612410.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
