Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM127C Antibody (OTI1A6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N271867A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271867A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271867A647 Supplier Novus Biologicals Supplier No. NBP271867AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM127C Monoclonal specifically detects FAM127C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen FAM127C
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1A6
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CXX1c, Family With Sequence Similarity 127 Member C, Family With Sequence Similarity 127, Member C, Mammalian Retrotransposon Derived Protein 8B, Mammalian Retrotransposon-Derived 2, MAR8B, Protein FAM127C, Retrotransposon Gag Like 8B, Retrotransposon Gag-Like Protein 8B, SIRH4, Sushi-Ichi Retrotransposon Homolog 1
Gene Symbols RTL8B
Host Species Mouse
Immunogen Full length recombinant protein of human FAM127C (NP_001071641) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 441518.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.