Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM127C Antibody (OTI1A6) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271867 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271867 Supplier Novus Biologicals Supplier No. NBP271867
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM127C Monoclonal specifically detects FAM127C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen FAM127C
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone OTI1A6
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias CXX1c, Family With Sequence Similarity 127 Member C, Family With Sequence Similarity 127, Member C, Mammalian Retrotransposon Derived Protein 8B, Mammalian Retrotransposon-Derived 2, MAR8B, Protein FAM127C, Retrotransposon Gag Like 8B, Retrotransposon Gag-Like Protein 8B, SIRH4, Sushi-Ichi Retrotransposon Homolog 1
Gene Symbols l(2)gl
Host Species Mouse
Immunogen Full length human recombinant protein of human FAM127C (NP_001071641) produced in HEK293T cell.
Molecular Weight of Antigen 13.1 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 441518
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.