Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM128B Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095428 20 μL
NB095427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095428 Supplier Novus Biologicals Supplier No. NBP30329120UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM128B Polyclonal specifically detects FAM128B in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM128B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FAM128Bmember B, mitotic spindle organizing protein 2B, Mitotic-spindle organizing protein associated with a ring of gamma-tubulin 2B, MOZART2BMGC87017
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-158 of human FAM128B (NP_079305.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80097
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.