Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM128B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FAM128B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FAM128B Polyclonal specifically detects FAM128B in Human samples. It is validated for Western Blot.Specifications
FAM128B | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
80097 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
FAM128Bmember B, mitotic spindle organizing protein 2B, Mitotic-spindle organizing protein associated with a ring of gamma-tubulin 2B, MOZART2BMGC87017 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-158 of human FAM128B (NP_079305.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title