Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM12B Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB030516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB030516 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB030516 Supplier Novus Biologicals Supplier No. NBP2825870.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM12B Polyclonal specifically detects FAM12B in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM12B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias epididymal protein 3B, epididymal secretory protein E3 beta, epididymal secretory protein E3-beta, FAM12B, HE3B, HE3-BETA, human epididymis-specific 3 beta, human epididymis-specific protein 3-beta, member B (epididymal), UNQ6412/PRO21187
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human FAM12B
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64184
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.