Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM130A1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106085 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106085 Supplier Novus Biologicals Supplier No. NBP106085
Add to Cart
Add to Cart

Goat Polyclonal Antibody

FAM130A1 Polyclonal specifically detects FAM130A1 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen FAM130A1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_700456.1
Gene Alias C12ORF2, C12orf22, CSRNP-2, cysteine/serine-rich nuclear protein 2, cysteine-serine-rich nuclear protein 2, FAM130A1, family with sequence similarity 130, member A1, FLJ25576, Protein FAM130A1, TAIP12, TAIP-12chromosome 12 open reading frame 22, TGF-beta-induced apoptosis protein 12
Gene Symbols CSRNP2
Host Species Goat
Immunogen Peptide with sequence C-KRQKQLRRKNVRFDQ corresponding to internal region according to Uniprot Mouse NP_700456.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81566
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.