Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM132B Antibody, Novus Biologicals™
SDP

Catalog No. NB393805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393805 25 μL
NBP257732 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393805 Supplier Novus Biologicals Supplier No. NBP25773225UL

Rabbit Polyclonal Antibody has been used in 2 publications

FAM132B Polyclonal specifically detects FAM132B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen FAM132B
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 -0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunoprecipitation
Formulation PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Gene Alias C1qTNF15, Complement C1q Tumor Necrosis Factor-Related Protein 15, CTRP15, ERFE, Erythroferrone, Family With Sequence Similarity 132, Member B, Myonectin, Protein FAM132B
Gene Symbols FAM132B
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EPTAERAHSVDPRDAWMLFVRQSDKGVNGKKRSRGKAKKLKFGLPGP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151176
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.