Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM134A Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP276379 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276379 Supplier Novus Biologicals Supplier No. NBP276379

Rabbit Polyclonal Antibody

FAM134A Polyclonal specifically detects FAM134A in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen FAM134A
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoprecipitation 15-20 μL/mg lysate
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias C2orf17, DKFZp686C2379, family with sequence similarity 134, member A, hypothetical protein LOC79137, MGC3035
Gene Symbols FAM134A
Host Species Rabbit
Immunogen Immunogen sequence between amino acids 235 and 375
Purification Method Antigen Affinity-purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 79137.0
Target Species Human
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.