Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM149B1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB135556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135556 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135556 Supplier Novus Biologicals Supplier No. NBP306311FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM149B1 Polyclonal specifically detects FAM149B1 in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM149B1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias family with sequence similarity 149, member B1, KIAA0974hypothetical protein LOC317662
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAM149B1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317662
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.