Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM153B Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB126443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB126443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB126443 Supplier Novus Biologicals Supplier No. NBP310771100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM153B Polyclonal specifically detects FAM153B in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM153B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias DKFZp434D115, family with sequence similarity 153, member B
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human FAM153B. Peptide sequence DPATLAKQLEDSTITGSHQQMSASPSSAPAEEATEKTKVEEEVKTRKPKK
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202134
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.