Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM155A Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB050956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050956 Supplier Novus Biologicals Supplier No. NBP298011AF350

Rabbit Polyclonal Antibody

FAM155A Polyclonal specifically detects FAM155A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAM155A
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias family with sequence similarity 155, member A
Gene Symbols NALF1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAM155A fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728215
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.