Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM161B Antibody, Novus Biologicals™
SDP

Catalog No. NBP15672220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15672220 20 μL
NBP156722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15672220 Supplier Novus Biologicals Supplier No. NBP15672220UL

Rabbit Polyclonal Antibody

FAM161B Polyclonal specifically detects FAM161B in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM161B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96MY7
Gene Alias c14_5547, C14orf44, chromosome 14 open reading frame 44, family with sequence similarity 161, member B, FLJ31697, hypothetical protein LOC145483
Gene Symbols FAM161B
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C14ORF44 The peptide sequence was selected from the middle region of C14ORF44. Peptide sequence AMDPHKSLEEVFKAKLKENRNNDRKRAKEYKKELEEMKQRIQTRPYLFEQ.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145483
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.