Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM162A Antibody, Novus Biologicals™
SDP

Catalog No. NBP258663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP258663 100 μL
NB435284 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP258663 Supplier Novus Biologicals Supplier No. NBP258663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM162A Polyclonal specifically detects FAM162A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAM162A
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Gene Alias chromosome 3 open reading frame 28, E2IG5C3orf28growth and transformation-dependent protein, E2-induced gene 5 protein, family with sequence similarity 162, member A, HGTD-P, HIF-1 alpha-responsive proapoptotic molecule
Gene Symbols FAM162A
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EGKKAAQRHETLTSLNLEKKARLKEEAAMKAK
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 26355
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.