Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM162A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | FAM162A |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
FAM162A Polyclonal specifically detects FAM162A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
FAM162A | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
26355 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EGKKAAQRHETLTSLNLEKKARLKEEAAMKAK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse | |
chromosome 3 open reading frame 28, E2IG5C3orf28growth and transformation-dependent protein, E2-induced gene 5 protein, family with sequence similarity 162, member A, HGTD-P, HIF-1 alpha-responsive proapoptotic molecule | |
FAM162A | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title