Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM163B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | FAM163B |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
FAM163B Polyclonal specifically detects FAM163B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FAM163B | |
Unconjugated | |
RUO | |
C9orf166, Chromosome 9 Open Reading Frame 166, Family With Sequence Similarity 163, Member B, Protein FAM163B | |
FAM163B | |
IgG | |
Affinity Purified |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
642968 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EEEPDFAVHSHLPPLHSNRNLVLTNGPALYPTASTSFSQKSPQARALCRSCSHCEPPTFFLQEPPEEEEDVLNGGERVLYKSVSQE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title