Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM177A1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290847 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290847 Supplier Novus Biologicals Supplier No. NBP178754PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM177A1 Polyclonal antibody specifically detects FAM177A1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen FAM177A1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias C14orf24, chromosome 14 open reading frame 24, DKFZp686J1254, family with sequence similarity 177, member A1, FLJ38854, hypothetical protein LOC283635
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Family with Sequence Similarity 177, Member A1 using the numbering given in entry NP_001072987.1 (GeneID 283635).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 283635
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.