Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM188A Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB000934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000934 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000934 Supplier Novus Biologicals Supplier No. NB1002504SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM188A Polyclonal specifically detects FAM188A in Human samples. It is validated for ELISA.

Specifications

Antigen FAM188A
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias C10orf97caspase recruitment domain containing pro-apoptotic protein, CARPRP11-394I23.1, chromosome 10 open reading frame 97, Dermal papilla-derived protein 5, DERP5CARD-containing protein, family with sequence similarity 188, member A, FLJ13397, MST126, my042, Protein CARP
Gene Symbols FAM188A
Host Species Rabbit
Immunogen Residues 59-76 AFLLKKLLFSSEKSSWRD of human CARP protein.
Purification Method Affinity Purified
Quantity 0.025 mL
Primary or Secondary Primary
Gene ID (Entrez) 80013
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.