Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM200A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | FAM200A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16007720
![]() |
Novus Biologicals
NBP16007720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP160077
![]() |
Novus Biologicals
NBP160077 |
100 μL |
Each for $487.50
|
|
|||||
Description
FAM200A Polyclonal specifically detects FAM200A in Human samples. It is validated for Western Blot.Specifications
FAM200A | |
Polyclonal | |
Rabbit | |
Q8TCP9 | |
221786 | |
Synthetic peptides corresponding to FAM200A The peptide sequence was selected from the middle region of FAM200A. Peptide sequence QTFNYYFPEEKFESLKENIWMKDPFAFQNPESIIELNLEPEEENELLQLS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C7orf38, chromosome 7 open reading frame 38, DKFZp727G131, family with sequence similarity 200, member A, FLJ36794, hypothetical protein LOC221786 | |
FAM200A | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title