Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM20C Antibody, Novus Biologicals™
SDP

Catalog No. NBP17076820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17076820 20 μL
NBP170768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17076820 Supplier Novus Biologicals Supplier No. NBP17076820UL

Rabbit Polyclonal Antibody

FAM20C Polyclonal specifically detects FAM20C in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen FAM20C
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Alias dentin matrix protein 4, DKFZp547C074, DMP4, DMP-4, family with sequence similarity 20, member C, RNS
Gene Symbols FAM20C
Host Species Rabbit
Immunogen Synthetic peptides corresponding to FAM20C(family with sequence similarity 20, member C) The peptide sequence was selected from the C terminal of FAM20C. Peptide sequence NETFIIHLDNGRGFGKYSHDELSILVPLQQCCRIRKSTYLRLQLLAKEEY.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56975
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.