Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM213A Mouse anti-Human, Clone: 7-B7, Novus Biologicals™
SDP

Catalog No. NBP275486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275486 Supplier Novus Biologicals Supplier No. NBP275486

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

FAM213A Monoclonal specifically detects FAM213A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAM213A
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 7-B7
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias C10orf58, Chromosome 10 Open Reading Frame 58, Family With Sequence Similarity 213, Member A, PAMM, Peroxiredoxin (PRX)-Like 2 Activated In M-CSF Stimulated Monocytes, Peroxiredoxin-Like 2 Activated In M-CSF Stimulated Monocytes, Protein PAMM, Redox-Regulatory Protein PAMM, UPF0765 Protein C10orf58
Gene Symbols PRXL2A
Host Species Mouse
Immunogen Recombinant protein within Human FAM213A aa 1-229/ 229. (SwissProt: Q9BRX8 Human)
Molecular Weight of Antigen 26 kDa
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 84293.0
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.