Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM213B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191435
Description
FAM213B Polyclonal specifically detects FAM213B in Mouse samples. It is validated for Western Blot.Specifications
FAM213B | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
2810405K02Rik, C1orf93, chromosome 1 open reading frame 93, DKFZp547M123, family with sequence similarity 213, member B, prostamide/PG F synthase | |
Rabbit | |
22 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Mouse: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_079858 | |
PRXL2B | |
The specific Immunogen is proprietary information. Peptide sequence SKQIYKELGFKRYNSLSILPAALGKPVRDVASKAKAVGIQGNLSGDLLQS. | |
Affinity purified | |
RUO | |
127281 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction