Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM219B Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB293043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293043 Supplier Novus Biologicals Supplier No. NBP326092JF646

Rabbit Polyclonal Antibody

FAM219B Polyclonal antibody specifically detects FAM219B in Human samples. It is validated for Western Blot, Immunocytochemistry

Specifications

Antigen FAM219B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias C15orf17, chromosome 15 open reading frame 17, FLJ00005, hypothetical protein LOC57184
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAM219B fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57184
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.