Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM21A Antibody, Novus Biologicals™
SDP

Catalog No. NB393131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393131 25 μL
NBP259796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393131 Supplier Novus Biologicals Supplier No. NBP25979625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM21A Polyclonal specifically detects FAM21A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FAM21A
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide
Gene Alias Alternative Protein FAM21B, BA56A21.1, bA98I6.1, FAM21B, Family With Sequence Similarity 21 Member A, Family With Sequence Similarity 21, Member A, Family With Sequence Similarity 21, Member B, WASH Complex Subunit FAM21A, WASH Complex Subunit FAM21B
Gene Symbols WASHC2A
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acid sequence:VRVYDEEVEEPVLKAEAEKTEQEKTREQKEVDLIPKVQEAVNYGLQVLDSAFEQLDIKAGNSDSEEDDANGRVELILEPKDLYIDRPLPYLIGSKLFME
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387680
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.