Learn More
Description
Specifically detects FAM21C in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | FAM21C |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1:100-1:1000 |
| Formulation | PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | Em:AC012044.3, family with sequence similarity 21, member C, KIAA0592FAM21A, VPEFVaccinia virus penetration factor, WASH complex subunit FAM21C |
| Gene Symbols | FAM21C |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human FAM21C. The exact sequence is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
