Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM32A Rabbit anti-Human, DyLight 350, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB135888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135888 Supplier Novus Biologicals Supplier No. NBP306327UV

Rabbit Polyclonal Antibody

FAM32A Polyclonal specifically detects FAM32A in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM32A
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZP586O0120, family with sequence similarity 32, member A, hypothetical protein LOC26017
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAM32A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26017
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.