Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM3A Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB159718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159718 100 μg
NB159719 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159718 Supplier Novus Biologicals Supplier No. NBP317844100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM3A Polyclonal antibody specifically detects FAM3A in Human samples. It is validated for Immunofluorescence

Specifications

Antigen FAM3A
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias 2.19, Cytokine-like protein 2-19, DLD, DXS560S, family with sequence similarity 3, member A, member A protein
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: FPRIQQLFTSPESSVTAAPRARKYKCGLPQPCPEEHLAFRVVSGAANVIGPKICLEDKMLM
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60343
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.