Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM3C Antibody, Novus Biologicals™
SDP

Catalog No. NBP224464SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224464SS 0.025 mg
NBP224464 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224464SS Supplier Novus Biologicals Supplier No. NBP224464SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM3C Polyclonal specifically detects FAM3C in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen FAM3C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-3 ug/ml, Simple Western 1:20, Immunohistochemistry-Paraffin 5-10 ug/ml
Formulation Tris-Glycine with 0.02% Sodium Azide
Gene Accession No. Q92520
Gene Alias family with sequence similarity 3, member C, GS3876, ILEIInterleukin-like EMT inducer, predicted osteoblast protein
Gene Symbols FAM3C
Host Species Rabbit
Immunogen A portion of amino acids 40-80 of human FAM3C was used as the immunogen for this antibody.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10447
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human,monkey, cow, dog and chicken, 92% homologous in chimpanzee, 91% homologous in mouse and rat, 90% homologous in opossum and 88% homologous in pufferfish.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.