Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM3C Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$192.47 - $529.95

Specifications

Antigen FAM3C
Concentration 1.0 mg/mL
Dilution Western Blot 0.5-3 ug/ml, Simple Western 1:20, Immunohistochemistry-Paraffin 5-10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224464SS
SDP
View Documents
Novus Biologicals
NBP224464SS
0.025 mg
Each for $192.47
Only null left
Add to Cart
 
NBP224464
SDP
View Documents
Novus Biologicals
NBP224464
0.1 mg
Each for $529.95
Only null left
Add to Cart
 
Description

Description

FAM3C Polyclonal specifically detects FAM3C in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FAM3C
Western Blot 0.5-3 ug/ml, Simple Western 1:20, Immunohistochemistry-Paraffin 5-10 ug/ml
Polyclonal
Rabbit
Immunology
Tris-Glycine with 0.02% Sodium Azide
family with sequence similarity 3, member C, GS3876, ILEIInterleukin-like EMT inducer, predicted osteoblast protein
FAM3C
IgG
Affinity Purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human,monkey, cow, dog and chicken, 92% homologous in chimpanzee, 91% homologous in mouse and rat, 90% homologous in opossum and 88% homologous in pufferfish.
1.0 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
Q92520
10447
A portion of amino acids 40-80 of human FAM3C was used as the immunogen for this antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.