Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM3C Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB337471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337471 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337471 Supplier Novus Biologicals Supplier No. NBP224464PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM3C Polyclonal antibody specifically detects FAM3C in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FAM3C
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias family with sequence similarity 3, member C, GS3876, ILEIInterleukin-like EMT inducer, predicted osteoblast protein
Host Species Rabbit
Immunogen A portion of amino acids 40-80 of human FAM3C was used as the immunogen for this antibody.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10447
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.