Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM43B Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB144997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB144997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB144997 Supplier Novus Biologicals Supplier No. NBP305973AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM43B Polyclonal specifically detects FAM43B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FAM43B
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FAM43B family with sequence similarity 43, member B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FAM43B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163933
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.