Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM46A Antibody, Novus Biologicals™
SDP

Catalog No. NBP15548820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15548820 20 μL
NBP155488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15548820 Supplier Novus Biologicals Supplier No. NBP15548820UL

Rabbit Polyclonal Antibody

FAM46A Polyclonal specifically detects FAM46A in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM46A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q96IP4
Gene Alias C6orf37, chromosome 6 open reading frame 37, family with sequence similarity 46, member A, FLJ20037, FLJ31495, HBV XAg-transactivated protein 11, HBV X-transactivated gene 11 protein, hypothetical protein LOC55603, retinal expressed gene C6orf37, XTP11
Gene Symbols FAM46A
Host Species Rabbit
Immunogen Synthetic peptides corresponding to FAM46A(family with sequence similarity 46, member A) The peptide sequence was selected from the N terminal of FAM46A. Peptide sequence MAEGEGYFAMSEDELACSPYIPLGGDFGGGDFGGGDFGGGDFGGGGSFGG.
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55603
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.