Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM46A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | FAM46A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15548820
![]() |
Novus Biologicals
NBP15548820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155488
![]() |
Novus Biologicals
NBP155488 |
100 μL |
Each for $487.50
|
|
|||||
Description
FAM46A Polyclonal specifically detects FAM46A in Human samples. It is validated for Western Blot.Specifications
FAM46A | |
Polyclonal | |
Rabbit | |
Q96IP4 | |
55603 | |
IgG | |
50 kDa |
Western Blot | |
Unconjugated | |
RUO | |
C6orf37, chromosome 6 open reading frame 37, family with sequence similarity 46, member A, FLJ20037, FLJ31495, HBV XAg-transactivated protein 11, HBV X-transactivated gene 11 protein, hypothetical protein LOC55603, retinal expressed gene C6orf37, XTP11 | |
Synthetic peptides corresponding to FAM46A(family with sequence similarity 46, member A) The peptide sequence was selected from the N terminal of FAM46A. Peptide sequence MAEGEGYFAMSEDELACSPYIPLGGDFGGGDFGGGDFGGGDFGGGGSFGG. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title