Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM59A Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341815 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341815 Supplier Novus Biologicals Supplier No. NBP281933MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM59A Polyclonal antibody specifically detects FAM59A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen FAM59A
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Chromosome 18 Open Reading Frame 11, chromosome 18 open reading frame 11, family with sequence, FAM59A, Family With Sequence Similarity 59, Member A, FLJ21610, GAREM, Gm944, GRB2 associated, regulator of MAPK1, Grb2-Associated And Regulator Of Erk/MAPK, GRB2-Associated And Regulator Of MAPK Protein, GRB2-Associated And Regulator Of MAPK1, Protein FAM59A
Host Species Rabbit
Immunogen FAM59A antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human FAM59A. The immunogen is located within amino acids 630 - 680 of FAM59A. Amino Acid Squence: DPSRSTSTPRQKTPGTPKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 64762
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.