Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM72A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | FAM72A |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Rabbit |
Description
FAM72A Polyclonal specifically detects FAM72A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FAM72A | |
Unconjugated | |
Rabbit | |
Human | |
729533 | |
This antibody was developed against a recombinant protein corresponding to the amino acid sequence:CVSILCCKFCKQVLSSRGMKAVLLADTEIDLFSTDIPPTNAVDFTGRCYFT | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Polyclonal | |
Purified | |
RUO | |
Family With Sequence Similarity 72 Member A, Family With Sequence Similarity 72, Member A, Latent Membrane Protein 1-Induced Protein, LMP1-Induced Protein, LMPIP, Protein FAM72A, Ugene | |
FAM72A | |
IgG | |
Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title