Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM73A Antibody, Novus Biologicals™
SDP

Catalog No. NBP216420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216420 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216420 Supplier Novus Biologicals Supplier No. NBP216420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM73A Polyclonal specifically detects FAM73A in Human samples. It is validated for Western Blot.

Specifications

Antigen FAM73A
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q8NAN2
Gene Alias DKFZp686M07166, family with sequence similarity 73, member A
Gene Symbols FAM73A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human FAM73A. The exact sequence is proprietary.
Molecular Weight of Antigen 71 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374986
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (86%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.