Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM82A Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | FAM82A |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FAM82A Polyclonal specifically detects FAM82A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
FAM82A | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
151393 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FAM82A1. The immunogen is located within amino acids 140 - 190 of FAM82A1. Amino Acid Squence: ITANTDTEEQSFPVPK | |
Primary |
1 mg/mL | |
Polyclonal | |
Chicken | |
Human, Mouse, Rat | |
family with sequence similarity 82, member A1, member A, UNQ9371/PRO34163 | |
RMDN2 | |
IgY | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title