Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM83H Antibody, Novus Biologicals™
SDP

Catalog No. NBP232218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232218 Supplier Novus Biologicals Supplier No. NBP232218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM83H Polyclonal specifically detects FAM83H in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen FAM83H
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_940890.3
Gene Alias family with sequence similarity 83, member H, protein FAM83H
Gene Symbols FAM83H
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 350 to 400 of human Family with Sequence Similarity 83, Member H using the numbering given in entry NP_940890.3 (GeneID 286077).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286077
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.