Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM89B Antibody, Novus Biologicals™
SDP

Catalog No. NBP223612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223612 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223612 Supplier Novus Biologicals Supplier No. NBP223612

Rabbit Polyclonal Antibody

FAM89B Polyclonal specifically detects FAM89B in Mouse samples. It is validated for Western Blot.

Specifications

Antigen FAM89B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias family with sequence similarity 89, member B, mammary tumor virus receptor homolog 1, MTVR1
Gene Symbols FAM89B
Host Species Rabbit
Immunogen Reacts with residues 152-163 [DDEEPPDASLPPC] of the 20 kDa MMTV-R.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23625
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.