Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCB Antibody (M38P3E10), PerCP, Novus Biologicals™
SDP

Catalog No. NB309195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309195 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309195 Supplier Novus Biologicals Supplier No. NBP252406PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FANCB Monoclonal antibody specifically detects FANCB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FANCB
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M38P3E10
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.8.1, EC 3.6.3.14, FA2, FAAP90, FAAP95FAB, FACB, Fanconi anemia group B protein, Fanconi anemia, complementation group B, Fanconi anemia-associated polypeptide of 95 kDa, FLJ34064, Protein FACB
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide- DRRENIHPYR
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 2187
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.