Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Antibody (1290D), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB254808APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254808APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB254808APC Supplier Novus Biologicals Supplier No. NBP254808APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular), Knockout Validated.

Specifications

Antigen FANCD2
Applications Flow Cytometry, Immunoassay
Classification Monoclonal
Clone 1290D
Conjugate APC
Dilution Flow Cytometry, Flow (Intracellular), Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Gene Symbols FANCD2
Host Species Rabbit
Immunogen This FANCD2 Antibody (1290D) [Allophycocyanin] was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2177
Target Species Human
Content And Storage Store at 4C in the dark.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.