Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ FANCD2 Antibody Blocking Peptide
Shop All R&D Systems Products

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 2177 |
Species | Human |
Concentration | 1 mg/mL |
Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-502 |
Gene Alias | DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 |
Gene Symbol | FANCD2 |
Quantity | 0.1 mg |
Research Category | Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction