Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100182SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100182SS 0.025 mL
NB100182 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100182SS Supplier Novus Biologicals Supplier No. NB100182SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 178 publications

FANCD2 Polyclonal specifically detects FANCD2 in Avian, Canine, Human, Kangaroo, Mouse, Primate, Rat, Zebrafish samples. It is validated for Chemotaxis, ChIP Assay, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, RNA Inhibition, Simple Western, Western Blot.

Specifications

Antigen FANCD2
Applications Chemotaxis, ChIP Assay, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, RNA Inhibition, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9BXW9
Gene Alias DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Gene Symbols FANCD2
Host Species Rabbit
Immunogen This FANCD2 Antibody was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Molecular Weight of Antigen 164.1 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2177
Test Specificity This antibody is specific for FANCD2.
Target Species Avian, Mouse, Canine, Human, Kangaroo, Kangaroo, Mouse, Zebrafish, Primate, Rat, Zebrafish
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.