Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 178 publications

$342.00 - $584.50

Specifications

Antigen FANCD2
Concentration 1.0 mg/mL
Dilution Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Applications Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100182SS
SDP
View Documents
Novus Biologicals
NB100182SS
0.025 mL
Each for $342.00
Only null left
Add to Cart
 
NB100182
SDP
View Documents
Novus Biologicals
NB100182
0.05 mL
Each for $584.50
Only null left
Add to Cart
 
Description

Description

FANCD2 Polyclonal specifically detects FANCD2 in Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, RNA Inhibition, Knockout Validated, Knockdown Validated.
Specifications

Specifications

FANCD2
Western Blot 1:10000-1:20000, Simple Western 1:25, Chromatin Immunoprecipitation reported in scientific literature (PMID 28196964), Flow Cytometry, Immunohistochemistry 2.5-5.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5.0 ug/ml, Immunoblotting reported in multiple pieces of scientific literature, RNA Inhibition reported in scientific literature (PMID 27694619), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Polyclonal
Rabbit
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
PBS with 0.05% Sodium Azide
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
FANCD2
IgG
Affinity Purified
This antibody is specific for FANCD2.
1.0 mg/mL
Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Unconjugated
RUO
Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish
Q9BXW9
2177
This FANCD2 Antibody was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
164.1 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.