Learn More
Description
Specifications
Specifications
| Antigen | FANCD2 |
| Applications | ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | FI-17 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 - 1:2000, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Formulation | Ascites with 0.1% Sodium Azide |
| Gene Accession No. | Q9BXW9 |
| Gene Alias | DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 |
| Gene Symbols | FANCD2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
