Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Antibody (FI-17), Novus Biologicals™
SDP

Catalog No. NB100316SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100316SS 0.025 mL
NB100316 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100316SS Supplier Novus Biologicals Supplier No. NB100316SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 13 publications

FANCD2 Monoclonal specifically detects FANCD2 in Hamster (Negative), Human, Mouse, Mouse (Negative) samples. It is validated for ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen FANCD2
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Simple Western, Western Blot
Classification Monoclonal
Clone FI-17
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. Q9BXW9
Gene Alias DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Gene Symbols FANCD2
Host Species Mouse
Immunogen This FANCD2 Antibody (FI-17) was developed against human FANCD2 fusion protein [Swiss-Prot: Q9BXW9]
Molecular Weight of Antigen 164.1 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2177
Test Specificity Human. This antibody does not react with mouse or hamster.
Target Species Hamster (Negative), Human, Mouse, Mouse (Negative)
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.