Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2, p Ser222 Antibody, Novus Biologicals™
SDP

Catalog No. NB100502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100502 0.2 mL
NB100502SS 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100502 Supplier Novus Biologicals Supplier No. NB100502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

FANCD2 (p Ser222) Polyclonal specifically detects FANCD2 (p Ser222) in Human samples. It is validated for Western Blot.

Specifications

Antigen FANCD2 (p Ser222)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:500
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. Q9BXW9
Gene Alias DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Gene Symbols FANCD2
Host Species Rabbit
Immunogen This FANCD2 [p Ser222] Antibody was developed against a synthetic peptide made to human FANCD2 (residues 217-226), surrounding the phosphorylated Serine 222. [Swiss-Prot# Q9BXW9]
Molecular Weight of Antigen 164.1 kDa
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2177
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.