Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FANCD2, p Ser222 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | FANCD2 (p Ser222) |
---|---|
Dilution | Western Blot 1:100-1:500 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100502SS
![]() |
Novus Biologicals
NB100502SS |
0.05 mL |
Each for $206.50
|
|
|||||
NB100502
![]() |
Novus Biologicals
NB100502 |
0.2 mL |
Each for $499.50
|
|
|||||
Description
FANCD2 Polyclonal specifically detects FANCD2 in Human samples. It is validated for Western Blot.Specifications
FANCD2 (p Ser222) | |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
Whole antisera with 0.1% Sodium Azide | |
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
FANCD2 | |
IgG | |
Unpurified | |
164.1 kDa |
Western Blot 1:100-1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
Q9BXW9 | |
2177 | |
This FANCD2 [p Ser222] Antibody was developed against a synthetic peptide made to human FANCD2 (residues 217-226), surrounding the phosphorylated Serine 222. [Swiss-Prot# Q9BXW9] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title