Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCD2 Antibody (1290C) - BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP261930SS

 View more versions of this product

Catalog No. NBP261930SS


Only null left
Add to Cart

Description

Description

FANCD2 Monoclonal specifically detects FANCD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Specifications

Specifications

FANCD2
Monoclonal
1.0mg/mL
Western Blot 0.5μg/ml, Knockout Validated
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Rabbit
164.1 kDa
0.025 mg
Primary
Human, Mouse, Rat
Purified
KnockDown, Western Blot
1290C
Unconjugated
PBS with 0.02% Sodium Azide
FANCD2
This FANCD2 Antibody (1290C) was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Protein A or G purified
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
2177
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.