Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCE Antibody (L5-P1E6*G5), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP250400AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250400AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250400AG Supplier Novus Biologicals Supplier No. NBP250400AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FANCE Monoclonal specifically detects FANCE in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen FANCE
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone L5-P1E6*G5
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FACEFAE, Fanconi anemia group E protein, Fanconi anemia, complementation group E, Protein FACE
Gene Symbols FANCE
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - KAALKHLGP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2178
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.